siRNA Details
| Gene Symbol: Ttyh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053273.2 | NP_444503.2 | 11, 12 | 1343 | ||
| GenBank mRNA | AF329682.1 | - | 11, 12 | 1283 | ||
| AK152172.1 | - | 11, 12 | 1295 | |||
| AK154835.1 | - | 11, 12 | 1350 | |||
| AK155166.1 | - | 11, 12 | 1344 | |||
| AK168408.1 | - | 11, 12 | 1296 | |||
| AK184196.1 | - | 1, 2 | 41 | |||
| BC060740.1 | - | 11, 12 | 1286 | 4158813 | ||
| BY731753.1 | - | 7, 8 | 653 | |||

Ttyh2
Genomic Map