siRNA Details
| Gene Symbol: Afg3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054070.3 | NP_473411.2 | 10 | 1313 | ||
| GenBank mRNA | AF329695.1 | - | 10 | 1254 | ||
| AK006708.1 | - | 4 | 604 | |||
| AK159647.1 | - | 10 | 1277 | |||
| AK167964.1 | - | 10 | 1288 | |||
| AK168244.1 | - | 10 | 1287 | |||
| BC043320.1 | - | 1 | 135 | |||
| BC056978.1 | - | 10 | 1272 | 6821746 | ||
| BC060064.1 | - | 10 | 1272 | |||

Afg3l1
Genomic Map