siRNA Details
Gene Symbol: Alpk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054085.2 | NP_473426.2 | 6 | 2170 | ||
GenBank mRNA | AF338872.1 | - | 6 | 2164 | ||
AK220384.1 | - | 2 | 553 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Alpk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054085.2 | NP_473426.2 | 6 | 2170 | ||
GenBank mRNA | AF338872.1 | - | 6 | 2164 | ||
AK220384.1 | - | 2 | 553 |