siRNA Details
| Gene Symbol: HBD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000519.3 | NP_000510.1 | 2 | 404 | ||
| GenBank mRNA | AY034468.1 | - | 2 | 404 | ||
| BC069307.1 | - | 2 | 259 | 7262103 | ||
| BC070282.1 | - | 2 | 260 | 30418964 | ||
| BU664913.1 | - | 2 | 223 | |||

HBD
Genomic Map