siRNA Details
| Gene Symbol: Psmd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080554.2 | NP_542121.2 | 4, 5 | 599 | ||
| GenBank mRNA | AK005335.1 | - | 4, 5 | 577 | ||
| AK011026.1 | - | 4, 5 | 576 | |||
| AK029067.1 | - | 3, 4 | 896 | |||
| AK032992.1 | - | 4, 5 | 577 | |||
| AK078419.1 | - | 4, 5 | 577 | |||
| AK145587.1 | - | 4, 5 | 576 | |||
| AK148284.1 | - | 600 | ||||
| AK220321.1 | - | 4, 5 | 540 | |||
| BC019112.1 | - | 4, 5 | 610 | |||

Psmd5
Genomic Map