siRNA Details
| Gene Symbol: Tm9sf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080556.3 | NP_542123.3 | 4 | 542 | ||
| GenBank mRNA | AK010325.1 | - | 4 | 499 | ||
| AK030340.1 | - | 4 | 472 | |||
| AK048012.1 | - | 4 | 461 | |||
| AK050338.1 | - | 4 | 489 | |||
| AK075720.1 | - | 4 | 542 | |||
| AK088905.1 | - | 4 | 514 | |||
| AK159757.1 | - | 4 | 487 | |||
| AK206314.1 | - | 1 | 21 | |||
| BC003862.1 | - | 4 | 481 | 3488326 | ||

Tm9sf2
Genomic Map