siRNA Details
| Gene Symbol: Ubox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001255993.1 | NP_001242922.1 | 5 | 798 | ||
| NM_001255994.1 | NP_001242923.1 | 4 | 651 | |||
| NM_080562.5 | NP_542129.2 | 4 | 656 | |||
| GenBank mRNA | AF360997.1 | - | 4 | 586 | ||
| AK034114.1 | - | 4 | 605 | |||
| AK049276.1 | - | 3 | 422 | |||
| AK158052.1 | - | 5 | 756 | |||
| AK170024.1 | - | 5 | 2499 | |||
| AK170100.1 | - | 4 | 627 | |||
| AK171070.1 | - | 5 | 765 | |||
| BC025068.1 | - | 3 | 518 | 4242459 | ||

Ubox5
Genomic Map