siRNA Details
| Gene Symbol: Acss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080575.2 | NP_542142.1 | 1 | 346 | ||
| GenBank mRNA | AB046742.1 | - | 1 | 335 | ||
| AK088244.1 | - | 1 | 325 | |||
| AK146568.1 | - | 1 | 347 | |||
| AK158119.1 | - | 1 | 341 | |||
| AK172412.1 | - | 1 | 332 | |||
| AK172470.1 | - | 1 | 321 | |||
| AK173275.1 | - | 1 | 307 | |||
| BC092278.1 | - | 1 | 375 | 3710505 | ||

Acss1
Genomic Map