siRNA Details
| Gene Symbol: Recql5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130454.2 | NP_569721.1 | 1, 2 | 712 | ||
| GenBank mRNA | AB038773.1 | - | 1, 2 | 46 | ||
| AB059999.1 | - | 1, 2 | 226 | |||
| AK029060.1 | - | 1, 2 | 708 | |||
| AK083513.1 | - | 1, 2 | 708 | |||
| BC053086.1 | - | 1, 2 | 526 | |||
| BC059000.1 | - | 1, 2 | 686 | |||
| BC060070.1 | - | 1, 2 | 526 | |||
| BC066176.1 | - | 1, 2 | 435 | |||

Recql5
Genomic Map