siRNA Details
| Gene Symbol: Grin3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130455.2 | NP_569722.1 | 2 | 700 | ||
| GenBank mRNA | AF396649.1 | - | 2 | 709 | ||
| AY062435.1 | - | 2 | 700 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grin3b| Gene Symbol: Grin3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130455.2 | NP_569722.1 | 2 | 700 | ||
| GenBank mRNA | AF396649.1 | - | 2 | 709 | ||
| AY062435.1 | - | 2 | 700 | |||