siRNA Details
| Gene Symbol: Kirrel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170985.1 | NP_001164456.1 | 5 | 832 | ||
| NM_130867.3 | NP_570937.2 | 5 | 829 | |||
| GenBank mRNA | AF480411.1 | - | 4 | 467 | ||
| AK158381.1 | - | 5 | 786 | |||
| AY017368.1 | - | 4 | 467 | |||
| AY243095.1 | - | 4 | 467 | |||
| BC023765.1 | - | 5 | 645 | 5343720 | ||

Kirrel
Genomic Map