siRNA Details
| Gene Symbol: Krtap19-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130870.1 | NP_570940.1 | 1 | 469 | ||
| GenBank mRNA | AF345291.1 | - | 1 | 469 | ||
| AK137740.1 | - | 1 | 165 | |||
| BC115787.1 | - | 1 | 466 | 40105094 | ||
| BC115815.1 | - | 1 | 469 | 40105095 | ||

Krtap19-2
Genomic Map