siRNA Details
| Gene Symbol: Krtap19-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130873.1 | NP_570943.1 | 1 | 221 | ||
| GenBank mRNA | AF345294.1 | - | 1 | 221 | ||
| BC116200.1 | - | 1 | 221 | 40090087 | ||
| BC116201.1 | - | 1 | 221 | 40090088 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap19-4| Gene Symbol: Krtap19-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130873.1 | NP_570943.1 | 1 | 221 | ||
| GenBank mRNA | AF345294.1 | - | 1 | 221 | ||
| BC116200.1 | - | 1 | 221 | 40090087 | ||
| BC116201.1 | - | 1 | 221 | 40090088 | ||