siRNA Details
| Gene Symbol: Cyp4f13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130882.1 | NP_570952.1 | 3 | 271 | ||
| GenBank mRNA | AF233643.1 | - | 3 | 271 | ||
| AK085008.1 | - | 3 | 302 | |||
| BC003954.1 | - | 3 | 280 | 3487830 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp4f13| Gene Symbol: Cyp4f13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130882.1 | NP_570952.1 | 3 | 271 | ||
| GenBank mRNA | AF233643.1 | - | 3 | 271 | ||
| AK085008.1 | - | 3 | 302 | |||
| BC003954.1 | - | 3 | 280 | 3487830 | ||