siRNA Details
Gene Symbol: Nxf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130888.1 | NP_570958.1 | 3 | 97 | ||
GenBank mRNA | AB178901.1 | - | 4 | 236 | ||
AY260550.1 | - | 3 | 97 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nxf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130888.1 | NP_570958.1 | 3 | 97 | ||
GenBank mRNA | AB178901.1 | - | 4 | 236 | ||
AY260550.1 | - | 3 | 97 |