siRNA Details
| Gene Symbol: Rnf144a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081977.2 | NP_001075446.1 | 6, 7 | 740 | ||
| NM_080563.4 | NP_542130.1 | 5, 6 | 666 | |||
| GenBank mRNA | AF360998.1 | - | 6, 7 | 736 | ||
| AK038586.1 | - | 5, 6 | 558 | |||
| AK133544.1 | - | 3, 4 | 280 | |||
| AK172904.1 | - | 4, 5 | 501 | |||
| BC030187.1 | - | 5, 6 | 630 | 5345434 | ||
| BQ884505.1 | - | 5, 6 | 599 | |||
| BU055293.1 | - | 7, 8 | 677 | |||

Rnf144a
Genomic Map