siRNA Details
| Gene Symbol: Emid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080595.2 | NP_542162.1 | 13 | 1104 | ||
| GenBank mRNA | AJ416093.1 | - | 13 | 1104 | ||
| AK153702.1 | - | 14 | 1223 | |||
| BC012248.1 | - | 4 | 201 | |||
| BC021849.1 | - | 3 | 103 | |||
| BC130023.1 | - | 13 | 1109 | 40140754 | ||

Emid1
Genomic Map