siRNA Details
| Gene Symbol: Ttbk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024856.2 | NP_001020027.1 | 4 | 704 | ||
| NM_001024857.2 | NP_001020028.1 | 3 | 630 | |||
| NM_080788.3 | NP_542966.2 | 4 | 458 | |||
| GenBank mRNA | AB046593.2 | - | 3 | 382 | ||
| AK046978.1 | - | 4 | 677 | |||
| AK053820.1 | - | 4 | 693 | |||
| AK077011.1 | - | 4 | 479 | |||
| AK137024.1 | - | 3 | 619 | |||
| AK148269.1 | - | 3 | 634 | |||
| BY741386.1 | - | 3 | 634 | |||

Ttbk2
Genomic Map