siRNA Details
| Gene Symbol: GDF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000557.4 | NP_000548.2 | 1 | 543 | ||
| NM_001319138.1 | NP_001306067.1 | 3 | 785 | |||
| XM_011529075.2 | XP_011527377.1 | 3 | 675 | |||
| GenBank mRNA | AB593125.1 | - | 1 | 531 | ||
| BC032495.1 | - | 3 | 785 | 5243109 | ||
| U13660.1 | - | 1 | 543 | |||

GDF5
Genomic Map