siRNA Details
| Gene Symbol: CD53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000560.3 | NP_000551.1 | 3 | 402 | ||
| NM_001040033.1 | NP_001035122.1 | 4 | 407 | |||
| NM_001320638.1 | NP_001307567.1 | 3 | 402 | |||
| GenBank mRNA | AK298723.1 | - | 3 | 349 | ||
| AK313582.1 | - | 3 | 355 | |||
| BC035456.1 | - | 4 | 389 | |||
| BC040693.1 | - | 3 | 332 | 5498854 | ||
| BG536920.1 | - | 3 | 403 | |||
| BT019643.1 | - | 2 | 236 | |||
| CA453414.1 | - | 409 | ||||
| M37033.1 | - | 3 | 329 | |||
| M60871.1 | - | 3 | 309 | |||

CD53
Genomic Map