siRNA Details
| Gene Symbol: Krtap19-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130857.2 | NP_570927.1 | 1 | 349 | ||
| GenBank mRNA | AF345295.1 | - | 1 | 301 | ||
| AK020697.1 | - | 1 | 350 | |||
| BC115545.2 | - | 1 | 301 | 40101652 | ||
| BC115546.2 | - | 1 | 301 | 40101653 | ||

Krtap19-3
Genomic Map