siRNA Details
| Gene Symbol: Baiap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037754.3 | NP_001032843.1 | 5, 6 | 452 | ||
| NM_001037755.3 | NP_001032844.2 | 5, 6 | 452 | |||
| NM_130862.4 | NP_570932.2 | 5, 6 | 452 | |||
| GenBank mRNA | AF390178.1 | - | 5, 6 | 410 | ||
| AF390179.1 | - | 5, 6 | 410 | |||
| AK049469.1 | - | 5, 6 | 453 | |||
| AK145924.1 | - | 5, 6 | 453 | |||
| AK158579.1 | - | 5, 6 | 477 | |||
| AK160401.1 | - | 5, 6 | 453 | |||
| BC006620.1 | - | 5, 6 | 443 | 3158004 | ||
| BC016411.1 | - | 5, 6 | 443 | 3158004 | ||

Baiap2
Genomic Map