siRNA Details
| Gene Symbol: Card14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130886.3 | NP_570956.1 | 4 | 573 | ||
| GenBank mRNA | AF363457.1 | - | 4 | 552 | ||
| AK086176.1 | - | 5 | 822 | |||
| BC029102.1 | - | 4 | 573 | 3979883 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Card14| Gene Symbol: Card14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130886.3 | NP_570956.1 | 4 | 573 | ||
| GenBank mRNA | AF363457.1 | - | 4 | 552 | ||
| AK086176.1 | - | 5 | 822 | |||
| BC029102.1 | - | 4 | 573 | 3979883 | ||