siRNA Details
| Gene Symbol: Nxf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130888.1 | NP_570958.1 | 4 | 269 | ||
| GenBank mRNA | AB178901.1 | - | 5 | 408 | ||
| AY260550.1 | - | 4 | 269 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nxf7| Gene Symbol: Nxf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130888.1 | NP_570958.1 | 4 | 269 | ||
| GenBank mRNA | AB178901.1 | - | 5 | 408 | ||
| AY260550.1 | - | 4 | 269 | |||