siRNA Details
Gene Symbol: Hps3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146323.1 | NP_001139795.1 | 2, 3 | 366 | ||
NM_001146324.1 | NP_001139796.1 | 2, 3 | 762 | |||
NM_080634.4 | NP_542365.3 | 2, 3 | 762 | |||
GenBank mRNA | AF393747.1 | - | 2, 3 | 751 | ||
AF393780.1 | - | 2, 3 | 754 | |||
AK034377.1 | - | 2, 3 | 733 | |||
AK036663.1 | - | 2, 3 | 770 | |||
AK037829.1 | - | 2, 3 | 261 | |||
AK049204.1 | - | 2, 3 | 755 | |||
AK079896.1 | - | 2, 3 | 754 | |||
AK086167.1 | - | 2, 3 | 751 | |||
AK149359.1 | - | 2, 3 | 748 | |||
AK155497.1 | - | 2, 3 | 729 | |||
AK157953.1 | - | 2, 3 | 747 | |||
AK166194.1 | - | 2, 3 | 747 | |||
AK169855.1 | - | 2, 3 | 733 | |||
AK170581.1 | - | 2, 3 | 278 | |||
AK171997.1 | - | 2, 3 | 354 | |||
BB660133.1 | - | 2, 3 | 359 | |||
BC058235.1 | - | 2, 3 | 736 | 6307400 |
