siRNA Details
| Gene Symbol: Cntnap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130457.2 | NP_569724.2 | 24 | 4060 | ||
| GenBank mRNA | AF333770.1 | - | 24 | 4025 | ||
| AK053611.1 | - | 22 | 3662 | |||
| AK142816.1 | - | 24 | 4056 | |||
| AK173255.1 | - | 24 | 4035 | |||
| BC034628.1 | - | 24 | 4060 | 5369648 | ||

Cntnap4
Genomic Map