siRNA Details
| Gene Symbol: Krtap19-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130857.2 | NP_570927.1 | 1 | 490 | ||
| GenBank mRNA | AF345295.1 | - | 1 | 442 | ||
| AK020697.1 | - | 1 | 491 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap19-3| Gene Symbol: Krtap19-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130857.2 | NP_570927.1 | 1 | 490 | ||
| GenBank mRNA | AF345295.1 | - | 1 | 442 | ||
| AK020697.1 | - | 1 | 491 | |||