siRNA Details
| Gene Symbol: Cd209e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130905.2 | NP_570975.1 | 3 | 224 | ||
| GenBank mRNA | AF373412.1 | - | 3 | 224 | ||
| AK040917.1 | - | 3 | 246 | |||
| BC107188.2 | - | 3 | 224 | 40050810 | ||
| BC107189.2 | - | 3 | 224 | 40050811 | ||

Cd209e
Genomic Map