siRNA Details
| Gene Symbol: C7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000587.2 | NP_000578.2 | 4, 5 | 391 | ||
| GenBank mRNA | AK290349.1 | - | 4, 5 | 401 | ||
| AK304848.1 | - | 4, 5 | 344 | |||
| AK312737.1 | - | 4, 5 | 325 | |||
| BC025402.1 | - | 4, 5 | 470 | |||
| BC063851.1 | - | 4, 5 | 372 | 6184213 | ||
| J03507.1 | - | 3, 4 | 277 | |||

C7
Genomic Map