siRNA Details
| Gene Symbol: Zfp287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133208.2 | NP_573471.2 | 1 | 657 | ||
| GenBank mRNA | AF281141.1 | - | 1 | 657 | ||
| AK029572.1 | - | 1 | 237 | |||
| AK031950.1 | - | 1 | 2508 | |||
| AK046006.1 | - | 2 | 689 | |||
| AK133603.1 | - | 2 | 670 | |||
| AK134983.1 | - | 1 | 235 | |||
| BC117876.1 | - | 1 | 382 | 40095930 | ||
| BC117877.1 | - | 1 | 382 | 40095933 | ||

Zfp287
Genomic Map