siRNA Details
| Gene Symbol: Sertad3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133210.2 | NP_573473.1 | 2 | 207 | ||
| GenBank mRNA | AF317202.1 | - | 2 | 211 | ||
| AK134829.1 | - | 2 | 280 | |||
| BC034886.1 | - | 2 | 207 | 1364881 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sertad3| Gene Symbol: Sertad3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133210.2 | NP_573473.1 | 2 | 207 | ||
| GenBank mRNA | AF317202.1 | - | 2 | 211 | ||
| AK134829.1 | - | 2 | 280 | |||
| BC034886.1 | - | 2 | 207 | 1364881 | ||