siRNA Details
| Gene Symbol: Tlr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290755.1 | NP_001277684.1 | 5 | 820 | ||
| NM_001290756.1 | NP_001277685.1 | 4 | 739 | |||
| NM_001290757.1 | NP_001277686.1 | 3 | 707 | |||
| NM_001290758.1 | NP_001277687.1 | 4 | 752 | |||
| NM_133211.4 | NP_573474.1 | 3 | 671 | |||
| GenBank mRNA | AF334942.1 | - | 3 | 614 | ||
| AF334943.1 | - | 3 | 628 | |||
| AK036549.1 | - | 3 | 629 | |||
| AK154906.1 | - | 5 | 778 | |||
| AK156908.1 | - | 3 | 675 | |||
| AK172680.1 | - | 1 | 4631 | |||
| AY035889.1 | - | 2 | 488 | |||
| BC120596.1 | - | 2 | 533 | 40129519 | ||
| BC132385.1 | - | 3 | 601 | 40130662 | ||

Tlr7
Genomic Map