siRNA Details
| Gene Symbol: Xpnpep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133216.3 | NP_573479.3 | 3 | 330 | ||
| GenBank mRNA | AF363970.1 | - | 3 | 270 | ||
| AK146033.1 | - | 3 | 310 | |||
| AK149676.1 | - | 3 | 269 | |||
| AK166715.1 | - | 3 | 230 | |||
| AK169024.1 | - | 2 | 156 | |||
| AK179231.1 | - | 1 | 27 | |||
| AK188589.1 | - | 1 | 27 | |||
| BC065174.1 | - | 3 | 218 | 6415314 | ||
| CJ081210.1 | - | 3 | 331 | |||

Xpnpep1
Genomic Map