siRNA Details
| Gene Symbol: Rfxap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133231.2 | NP_573494.1 | 1 | 517 | ||
| GenBank mRNA | AF335512.1 | - | 1 | 497 | ||
| AK087489.1 | - | 1 | 518 | |||
| AK144633.1 | - | 1 | 504 | |||
| BC019935.1 | - | 1 | 480 | 5041039 | ||
| BC047123.1 | - | 1 | 480 | |||

Rfxap
Genomic Map