siRNA Details
| Gene Symbol: Glcci1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286728.1 | NP_001273657.1 | 2 | 59 | ||
| NM_001286729.1 | NP_001273658.1 | 2 | 59 | |||
| NM_133236.3 | NP_573499.2 | 2 | 735 | |||
| GenBank mRNA | AK029524.1 | - | 2 | 61 | ||
| AY098636.1 | - | 2 | 781 | |||
| BC050802.1 | - | 2 | 97 | 6771009 | ||
| CA561411.1 | - | 2 | 115 | |||
| CN720421.1 | - | 2 | 92 | |||

Glcci1
Genomic Map