siRNA Details
| Gene Symbol: Cd209a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133238.5 | NP_573501.1 | 2 | 163 | ||
| GenBank mRNA | AF373408.1 | - | 2 | 110 | ||
| AF374470.1 | - | 2 | 110 | |||
| AK054139.1 | - | 2 | 127 | |||
| AK171639.1 | - | 2 | 107 | |||
| AY049062.1 | - | 2 | 163 | |||
| BC111095.1 | - | 1 | 211 | 40040036 | ||

Cd209a
Genomic Map