siRNA Details
| Gene Symbol: Ms4a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133246.5 | NP_573509.1 | 3 | 386 | ||
| GenBank mRNA | AF280401.1 | - | 3 | 325 | ||
| AF321128.1 | - | 3 | 325 | |||
| AK036464.1 | - | 3 | 388 | |||
| AK158372.1 | - | 3 | 387 | |||
| AY184359.1 | - | 3 | 340 | |||
| BC117013.1 | - | 3 | 302 | 40126332 | ||
| BC117015.1 | - | 3 | 302 | 40126334 | ||
| BC144853.1 | - | 3 | 302 | 9053401 | ||

Ms4a3
Genomic Map