siRNA Details
| Gene Symbol: Zfand2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159908.1 | NP_001153380.1 | 4, 5 | 533 | ||
| NM_133349.3 | NP_579927.1 | 4, 5 | 544 | |||
| GenBank mRNA | AF224494.1 | - | 4, 5 | 430 | ||
| AK146082.1 | - | 4, 5 | 482 | |||
| AK171245.1 | - | 4, 5 | 484 | |||
| AK198290.1 | - | 2, 3 | 161 | |||
| BC058780.1 | - | 4, 5 | 470 | 6312202 | ||

Zfand2a
Genomic Map