siRNA Details
| Gene Symbol: Mat1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133653.3 | NP_598414.1 | 4 | 670 | ||
| GenBank mRNA | AK149542.1 | - | 3 | 421 | ||
| AK153832.1 | - | 3 | 424 | |||
| BC011211.1 | - | 3 | 408 | 4196928 | ||
| L13622.1 | - | 3 | 422 | |||

Mat1a
Genomic Map