siRNA Details
Gene Symbol: Crk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277219.1 | NP_001264148.1 | 2 | 430 | ||
NM_133656.5 | NP_598417.2 | 2 | 430 | |||
GenBank mRNA | AF239673.1 | - | 2 | 300 | ||
AK028179.1 | - | 2 | 407 | |||
AK028488.1 | - | 2 | 380 | |||
AK036905.1 | - | 2 | 381 | |||
AK052795.1 | - | 2 | 389 | |||
AK076097.1 | - | 2 | 125 | |||
AK088519.1 | - | 2 | 401 | |||
AK139089.1 | - | 2 | 386 | |||
AK152549.1 | - | 2 | 404 | |||
AK154987.1 | - | 2 | 415 | |||
AK161356.1 | - | 2 | 380 | |||
AK161761.1 | - | 2 | 372 | |||
BC012216.1 | - | 2 | 366 | 3992341 | ||
BC031149.1 | - | 2 | 363 | 4975654 | ||
S72408.1 | - | 2 | 355 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
