siRNA Details
| Gene Symbol: Scml2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290651.1 | NP_001277580.1 | 4 | 832 | ||
| NM_001290652.1 | NP_001277581.1 | 4 | 832 | |||
| GenBank mRNA | AF285577.1 | - | 6 | 658 | ||
| AK016533.1 | - | 7 | 704 | |||
| AK031490.1 | - | 4 | 811 | |||
| AK040521.1 | - | 4 | 834 | |||
| BC056382.1 | - | 4 | 522 | 6853936 | ||

Scml2
Genomic Map