siRNA Details
| Gene Symbol: Gsx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133256.2 | NP_573555.1 | 2 | 968 | ||
| GenBank mRNA | AK159070.1 | - | 1 | 189 | ||
| BC119464.1 | - | 2 | 753 | |||
| BC119465.1 | - | 2 | 753 | |||
| BC134359.1 | - | 2 | 753 | |||
| S79041.1 | - | 2 | 968 | |||

Gsx2
Genomic Map