siRNA Details
| Gene Symbol: Prss8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133351.3 | NP_579929.1 | 4 | 517 | ||
| GenBank mRNA | AB038244.1 | - | 4 | 498 | ||
| AF188613.1 | - | 4 | 431 | |||
| AF378085.2 | - | 4 | 508 | |||
| AK010640.1 | - | 4 | 400 | |||
| AK078696.1 | - | 4 | 472 | |||
| AY335911.1 | - | 5 | 365 | |||
| BC003851.1 | - | 4 | 491 | 3600399 | ||

Prss8
Genomic Map