siRNA Details
| Gene Symbol: Zfp617 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133358.3 | NP_579936.2 | 2 | 266 | ||
| GenBank mRNA | AB020542.1 | - | 227 | |||
| BC014712.1 | - | 2 | 172 | 4507398 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp617| Gene Symbol: Zfp617 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133358.3 | NP_579936.2 | 2 | 266 | ||
| GenBank mRNA | AB020542.1 | - | 227 | |||
| BC014712.1 | - | 2 | 172 | 4507398 | ||