siRNA Details
| Gene Symbol: Krtap19-9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133359.2 | NP_579937.1 | 1 | 301 | ||
| GenBank mRNA | AK086935.1 | - | 1 | 303 | ||
| AY026312.1 | - | 1 | 302 | |||
| BC119613.1 | - | 1 | 290 | 40061693 | ||
| BC119614.1 | - | 1 | 290 | 40061695 | ||
| BC127239.1 | - | 1 | 290 | |||
| BC127240.1 | - | 1 | 290 | |||

Krtap19-9b
Genomic Map