siRNA Details
| Gene Symbol: Slc12a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133648.2 | NP_598409.2 | 4 | 339 | ||
| NM_133649.2 | NP_598410.2 | 4 | 1000 | |||
| GenBank mRNA | AF211854.1 | - | 4 | 495 | ||
| AF211855.1 | - | 4 | 339 | |||
| AK133006.1 | - | 4 | 1490 | |||
| AK137387.1 | - | 3 | 319 | |||
| BC062099.1 | - | 955 | 6844732 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001109630, XM_006234711, XM_006234710.

Slc12a6
Genomic Map