siRNA Details
| Gene Symbol: Xpnpep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289729.1 | NP_001276658.1 | 10 | 1582 | ||
| NM_133213.2 | NP_573476.2 | 10 | 1381 | |||
| GenBank mRNA | AF367247.1 | - | 10 | 1045 | ||
| AF428102.1 | - | 10 | 1232 | |||
| AK078835.1 | - | 10 | 1188 | |||
| AK085520.1 | - | 10 | 1380 | |||
| AK087627.1 | - | 10 | 1188 | |||
| AK164538.1 | - | 10 | 1209 | |||
| BC140977.1 | - | 10 | 1025 | 9055589 | ||

Xpnpep2
Genomic Map