siRNA Details
| Gene Symbol: Ripply3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB063284.1 | - | 4 | 769 | ||
| AK172430.1 | - | 4 | 760 | |||
| BC060291.1 | - | 4 | 725 | 30135244 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ripply3| Gene Symbol: Ripply3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB063284.1 | - | 4 | 769 | ||
| AK172430.1 | - | 4 | 760 | |||
| BC060291.1 | - | 4 | 725 | 30135244 | ||