siRNA Details
| Gene Symbol: Rfxap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133231.2 | NP_573494.1 | 3 | 946 | ||
| GenBank mRNA | AF335512.1 | - | 3 | 926 | ||
| AK077640.1 | - | 3 | 944 | |||
| AK087489.1 | - | 3 | 947 | |||
| AK144633.1 | - | 3 | 933 | |||
| BC019935.1 | - | 3 | 909 | 5041039 | ||
| BC047123.1 | - | 3 | 909 | |||
| BQ830204.1 | - | 1 | 205 | |||
| BX519947.1 | - | 1 | 263 | |||

Rfxap
Genomic Map