siRNA Details
| Gene Symbol: Prima1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133364.2 | NP_579942.2 | 5 | 819 | ||
| GenBank mRNA | AK021000.1 | - | 3 | 765 | ||
| AK039705.1 | - | 3 | 683 | |||
| AK140034.1 | - | 6 | 1082 | |||
| AY043275.1 | - | 5 | 821 | |||
| AY225515.1 | - | 6 | 992 | |||

Prima1
Genomic Map